Trimethylamine N-oxide dihydrate, 98+%
Trimethylamine N-oxide dihydrate, 98+%
Trimethylamine N-oxide dihydrate, 98+%
Thermo Scientific Chemicals

Trimethylamine N-oxide dihydrate, 98+%

CAS: 62637-93-8 | C3H13NO3 | 111.14 g/mol
数量:
5 g
25 g
100 g
500 g
製品番号(カタログ番号) A14916.22
または、製品番号A14916-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS62637-93-8
IUPAC NameN,N-dimethylmethanamine oxide dihydrate
Molecular FormulaC3H13NO3
InChI KeyPGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILESO.O.C[N+](C)(C)[O-]
さらに表示
Melting Point (clear melt)93.0-101.0°C
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Appearance (Color)White
Trimethylamine N-oxide dihydrate is used as an oxidizing agent in organic synthesis. It is also used to make quaternary ammonium compounds and a warning agent for flammable gas. Further, it acts as a flavoring agent or adjuvant. It serves as an oxidant for the catalytic osmium tetraoxide cis-hydroxylation of hindered olefins. In addition to this, it is used as a reactant for C-H bond cleavage and decarbonylating agent for solvent-free reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine N-oxide dihydrate is used as an oxidizing agent in organic synthesis. It is also used to make quaternary ammonium compounds and a warning agent for flammable gas. Further, it acts as a flavoring agent or adjuvant. It serves as an oxidant for the catalytic osmium tetraoxide cis-hydroxylation of hindered olefins. In addition to this, it is used as a reactant for C-H bond cleavage and decarbonylating agent for solvent-free reactions.

Solubility
Soluble in water, ethanol, dimethyl sulfoxide and methanol. Sparingly soluble in hot chloroform. Insoluble in diethyl ether, benzene and hydrocarbon solvents.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for the liberation of organics from their complexes with iron carbonyls. Partial oxidation of CO to CO2 takes place: J. Chem. Soc., Chem. Commun., 336 (1974).
  2. Also oxidizes alkylboranes to alcohols, giving less interference with other functionality than the more usual H2O2: J. Org. Chem., 40, 1776 (1975).
  3. Has been used in the oxidation of alkyl halides to aldehydes. For a procedure, including dehydration of the N-oxide by distillation of water with DMF, see: Org. Synth. Coll., 5, 872 (1973):
  4. For an improved procedure using DMSO, see: Tetrahedron Lett., 31, 4825 (1990).
  5. For use as a reoxidant in the catalytic cis-hydroxylation of alkenes with Osmium(VIII) oxide, 12103 , see: Tetrahedron Lett., 21, 449 (1980).
  6. Deprotonation by LDA at low temperature gives the unstable azomethine ylide, which undergoes 1,3-dipolar addition, even with unactivated alkenes, to give pyrrolidines: J. Chem. Soc., Chem. Commun., 31 (1983):
  7. Reviews: Trimethylamine as a useful oxidizing agent: J. Prakt. Chem./ Chem. Ztg., 338, 190 (1996); The utility of N-oxides in synthesis: Synthesis, 263 (1993).
  8. Compare also 4-Methyl morpholine N-oxide monohydrate, A15996 .
  9. Hunger, J.; Ottosson, N.; Mazur, K.; Bonn, M.; Bakker, H. J. Water-mediated interactions between trimethylamine-N-oxide and urea. Phys. Chem. Chem. Phys. 2015, 17 (1), 298-306.
  10. Makarov, D. M.; Egorov, G. I.; Kolker, A. M. Density and Volumetric Properties of Aqueous Solutions of Trimethylamine N-Oxide in the Temperature Range from (278.15 to 323.15) K and at Pressures up to 100 MPa. J. Chem. Eng. Data 2015, 60 (5), 1291-1299.