Acenaphthene, 97%
Acenaphthene, 97%
Acenaphthene, 97%
Thermo Scientific Chemicals

Acenaphthene, 97%

CAS: 83-32-9 | C12H10 | 154.21 g/mol
製品番号(カタログ番号) A14922.0B
または、製品番号A14922-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS83-32-9
IUPAC Name1,2-dihydroacenaphthylene
Molecular FormulaC12H10
InChI KeyCWRYPZZKDGJXCA-UHFFFAOYSA-N
SMILESC1CC2=C3C1=CC=CC3=CC=C2
さらに表示
FormTranslucent crystalline chunks, flakes and pale powder
Appearance (Color)White to cream or yellow to brown
Assay (GC)>96.0%
Acenaphthene is used in the preparation of naphthalene dicarboxylic anhydride, which is used as a precursor to dyes and optical brighteners. Further, it is used in the preparation of active pharmaceutical ingredients and in plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acenaphthene is used in the preparation of naphthalene dicarboxylic anhydride, which is used as a precursor to dyes and optical brighteners. Further, it is used in the preparation of active pharmaceutical ingredients and in plastics.

Solubility
Soluble in chloroform, benzene, tolune, glacial acetic acid and methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and alkaline earth metals.
RUO – Research Use Only

General References:

  1. Li, Z.; Liu, Y.; Yang, X.; Xing, Y.; Wang, Z.; Yang, Q.; Yang, R. T. Desorption Kinetics of Naphthalene and Acenaphthene over Two Activated Carbons via Thermogravimetric Analysis. Energy Fuels 2015, 29 (8), 5303-5310.
  2. Chen, Z.; Li, H.; Gao, Y.; Peng, A. Removal of phenanthrene and acenaphthene from aqueous solution by enzyme-catalyzed phenol coupling reaction. Chem. Eng. J. 2015, 265, 27-33.