4-Chloro-2-methylquinoline, 97%
4-Chloro-2-methylquinoline, 97%
4-Chloro-2-methylquinoline, 97%
Thermo Scientific Chemicals

4-Chloro-2-methylquinoline, 97%

CAS: 4295-06-1 | C10H8ClN | 177.63 g/mol
製品番号(カタログ番号) A15002.03
または、製品番号A15002-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4295-06-1
IUPAC Name4-chloro-2-methylquinoline
Molecular FormulaC10H8ClN
InChI KeyHQAIROMRVBVWSK-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=C2C=CC=CC2=N1
さらに表示
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.6195-1.6245 @ 20°C
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Abbas Alvanipour.; Lowell D. Kispert. Cobalt stearate-aluminum alkyl catalyzed hydrogenation of substituted quinolines, isoquinoline and naphthalene. Journal of Molecular Catalysis. 1988, 48, (2-3), 277-283.
  2. Shangdong Yan.; Bijie L.; Xiaobing Wa.; Zhangjie Sh. Ortho Arylation of Acetanilides via Pd(II)-Catalyzed C-H Functionalization. J. Am. Chem. Soc. 2007, 129, (19), 6066-6067.