Thermo Scientific Chemicals

Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A15008.14
25 g, Each
Thermo Scientific Chemicals

Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A15008.14
25 g, Each
Quantity
Catalog number: A15008.14
also known as A15008-14
Price (USD)
Price: 110.00
Online price: 94.65
Your price:
Quantity
-

Chemical Identifiers

CAS
622-78-6
IUPAC Name
(isothiocyanatomethyl)benzene
Molecular Formula
C8H7NS
InChI Key
MDKCFLQDBWCQCV-UHFFFAOYSA-N
SMILES
S=C=NCC1=CC=CC=C1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.6010-1.6050 @ 20?C

Description

Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoyl­acetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl isothiocyanate is used in a synthesis of 2-phenylpyrimidine-4-thiones from Benzoyl­acetonitrile. Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text