1-Bromohexadecane, 98%
1-Bromohexadecane, 98%
1-Bromohexadecane, 98%
Thermo Scientific Chemicals

1-Bromohexadecane, 98%

CAS: 112-82-3 | C16H33Br | 305.34 g/mol
数量:
25 g
250 g
1000 g
製品番号(カタログ番号) A15147.30
または、製品番号A15147-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS112-82-3
Appearance (Color)Colorless to white to brown
Assay (GC)≥97.5%
Refractive Index1.4605-1.4645 @ 20?C
FormFused solid or clear liquid as melt
Identification (FTIR)Conforms
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Pretima Manoj.; Andrew D Watson.; David J Hibberd.; Annette J Fillery-Travis.; Margaret M Robins. II. Steady-State Rheological Investigations ☆. Journal of Colloid and Interface Science. 1998, 207, (2), 294-302.
  2. Domenico Albanese.; Dario Landini.; Michele Penso. Hydrated Tetrabutylammonium Fluoride as a Powerful Nucleophilic Fluorinating Agent. J. Org. Chem. 1998, 63, (25), 9587-9589.