Methyl 3,5-dihydroxybenzoate, 98%
Methyl 3,5-dihydroxybenzoate, 98%
Methyl 3,5-dihydroxybenzoate, 98%
Methyl 3,5-dihydroxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 3,5-dihydroxybenzoate, 98%

CAS: 2150-44-9 | C8H8O4 | 168.15 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A15224.09
または、製品番号A15224-09
価格(JPY)
-
見積もりを依頼する
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2150-44-9
IUPAC Namemethyl 3,5-dihydroxybenzoate
Molecular FormulaC8H8O4
InChI KeyRNVFYQUEEMZKLR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(O)=CC(O)=C1
さらに表示
Appearance (Color)White to cream or pale brown
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)164.0-170.0?C
Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Michael S.Wendlan; Steven C.Zimmerman. Synthesis of cored dendrimers. J. Am. Chem. Soc. 1999, 121,(6), 1389-1390.
  2. Yadollah Delaviz; Harry W.Gibson. Macrocyclic polymers. 1. Synthesis of a poly(ester crown) based on bis(5-carboxy-1,3-phenylene)-32-crown-10 and 4,4'-isopropylidenediphenol (bisphenol A). Macromolecules. 1992, 25,(1), 18-20.