Thermo Scientific Chemicals

Phenyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals

Catalog number: A15326.30
250 g, Each
Thermo Scientific Chemicals

Phenyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals

Catalog number: A15326.30
250 g, Each
Quantity
Catalog number: A15326.30
also known as A15326-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
4207-56-1
IUPAC Name
N,N,N-trimethylanilinium; tribroman-2-uide
Molecular Formula
C9H14Br3N
InChI Key
PRXNKYBFWAWBNZ-UHFFFAOYSA-N
SMILES
Br[Br-]Br.C[N+](C)(C)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
110-118°C
Appearance (Color)
Orange
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%

Description

Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, reducing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text