1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
1-Methylnaphthalene, 96%
Thermo Scientific Chemicals

1-Methylnaphthalene, 96%

CAS: 90-12-0 | C11H10 | 142.201 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15327.14
または、製品番号A15327-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS90-12-0
IUPAC Name1-methylnaphthalene
Molecular FormulaC11H10
InChI KeyQPUYECUOLPXSFR-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=CC=C1
さらに表示
Appearance (Color)Clear colorless to yellow to orange to brown
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.6120-1.6180 @ 20?C
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

Solubility
Miscible with water, alcohol, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
RUO – Research Use Only

General References:

  1. Onyango, E. O.; Kelley, A. R.; Qian, D. C.; Gribble, G. W. Syntheses of 1-Bromo-8-methylnaphthalene and 1-Bromo-5-methylnaphthalene. J. Org. Chem. 2015, 80 (11), 5970-5972.
  2. Jaroszewska, K.; Masalska, A.; Grzechowiak, J. R.; Grams, J. Hydroconversion of 1-methylnaphthalene over Pt/AlSBA-15-Al2O3 composite catalysts. Appl. Catal., A 2015, 505, 116-130.