Methyl 3-hydroxy-4-nitrobenzoate, 98%
Methyl 3-hydroxy-4-nitrobenzoate, 98%
Methyl 3-hydroxy-4-nitrobenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-hydroxy-4-nitrobenzoate, 98%

CAS: 713-52-0 | C8H7NO5 | 197.146 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A15329.06
または、製品番号A15329-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS713-52-0
IUPAC Name5-(methoxycarbonyl)-2-nitrobenzen-1-olate
Molecular FormulaC8H6NO5
InChI KeyUEGCRFNWTGYVKX-UHFFFAOYSA-M
SMILESCOC(=O)C1=CC=C(C([O-])=C1)[N+]([O-])=O
さらに表示
FormCrystals or powder or crystalline powder or granules
Assay (GC)≥97.5%
Melting Point (clear melt)88.0-95.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only