Description
m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.
Solubility
Soluble in water, methanol, acetone, DMF, MEK, dioxane
Notes
Air & Light Sensitive. Store in the dark. Store under dry argon. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
RUO – Research Use Only