2,5-Diphenyl-p-benzoquinone, 96%
2,5-Diphenyl-p-benzoquinone, 96%
2,5-Diphenyl-p-benzoquinone, 96%
Thermo Scientific Chemicals

2,5-Diphenyl-p-benzoquinone, 96%

CAS: 844-51-9 | C18H12O2 | 260.29 g/mol
製品番号(カタログ番号) A15369.06
または、製品番号A15369-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS844-51-9
Assay (HPLC)≥95.0%
Melting Point (clear melt)213-221°C
Appearance (Color)Yellow to orange
FormPowder
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

Solubility
Soluble in hot Toluene (almost transparency).

Notes
Incompatible with Oxidizing agents, Reducing agent.
RUO – Research Use Only