Thermo Scientific Chemicals

Hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A15398.22
100 g, Each
Thermo Scientific Chemicals

Hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A15398.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15398.22
also known as A15398-22
Price (USD)
Price: 36.40
Online price: 30.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5470-11-1
IUPAC Name
hydroxylamine hydrochloride
Molecular Formula
ClH4NO
InChI Key
WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES
Cl.NO
Appearance (Color)
White
Form
Crystals or crystalline powder or powder
Comment
Sourced in UK and US
Identification (FTIR)
Conforms (UK sourced material only)
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (UK sourced material)

Description

Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

Solubility
Soluble in water, alcohol, methanol and glycerol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, phosphorous pentachloride, calcium and anhydrous copper(II) sulfate.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text