DL-2,4-Diaminobutyric acid dihydrochloride, 99%
DL-2,4-Diaminobutyric acid dihydrochloride, 99%
DL-2,4-Diaminobutyric acid dihydrochloride, 99%
DL-2,4-Diaminobutyric acid dihydrochloride, 99%
Thermo Scientific Chemicals

DL-2,4-Diaminobutyric acid dihydrochloride, 99%

CAS: 65427-54-5 | C4H12Cl2N2O2 | 191.052 g/mol
製品番号(カタログ番号) A15408.03
または、製品番号A15408-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS65427-54-5
IUPAC Name(2R)-2,4-diazaniumylbutanoate
Molecular FormulaC4H11N2O2
InChI KeyOGNSCSPNOLGXSM-GSVOUGTGSA-O
SMILES[NH3+]CC[C@@H]([NH3+])C([O-])=O
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activityagainst mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activity against mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

Solubility
Soluble in water 50 mg/mL.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Sandra M Leal; Wendi S Neckameyer. Pharmacological evidence for GABAergic regulation of specific behaviors in Drosophila melanogaster. Journal of Neurobiology. 2002, 50, (3), 245-261.
  2. Sandra M Leal; Nitin Kumar; Wendi S Neckameyer. GABAergic modulation of motor-driven behaviors in juvenile Drosophila and evidence for a nonbehavioral role for GABA transport. Journal of Neurobiology. 2004, 61(2), 189-208.