Thermo Scientific Chemicals

Chromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15438.14
25 g, Each
Thermo Scientific Chemicals

Chromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A15438.14
25 g, Each
Quantity
Catalog number: A15438.14
also known as A15438-14
Price (USD)
Price: 113.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17422-74-1
IUPAC Name
4-oxo-4H-chromene-3-carbaldehyde
Molecular Formula
C10H6O3
InChI Key
FSMYWBQIMDSGQP-UHFFFAOYSA-N
SMILES
O=CC1=COC2=CC=CC=C2C1=O
Assay (GC)
≥96.0%
Form
Powder
Appearance (Color)
Cream to yellow to pale orange
Melting Point (clear melt)
147.0-156.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text