Chromone-3-carboxaldehyde, 97%
Chromone-3-carboxaldehyde, 97%
Chromone-3-carboxaldehyde, 97%
Thermo Scientific Chemicals

Chromone-3-carboxaldehyde, 97%

CAS: 17422-74-1 | C10H6O3 | 174.155 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A15438.22
または、製品番号A15438-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS17422-74-1
IUPAC Name4-oxo-4H-chromene-3-carbaldehyde
Molecular FormulaC10H6O3
InChI KeyFSMYWBQIMDSGQP-UHFFFAOYSA-N
SMILESO=CC1=COC2=CC=CC=C2C1=O
さらに表示
Assay (GC)≥96.0%
FormPowder
Appearance (Color)Cream to yellow to pale orange
Melting Point (clear melt)147.0-156.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only