Thermo Scientific Chemicals

3-Dimethylaminopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A15459.22
100 g, Each
Thermo Scientific Chemicals

3-Dimethylaminopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A15459.22
100 g, Each
Quantity
Catalog number: A15459.22
also known as A15459-22
Price (USD)
Price: 38.50
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5407-04-5
IUPAC Name
hydrogen (3-chloropropyl)dimethylamine chloride
Molecular Formula
C5H13Cl2N
InChI Key
LJQNMDZRCXJETK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCCl
Appearance (Color)
White
Identification (FTIR)
Conforms
Melting Point (clear melt)
136.0-144.0°C
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤1%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text