2-Ethylfuran, 98+%
2-Ethylfuran, 98+%
2-Ethylfuran, 98+%
Thermo Scientific Chemicals

2-Ethylfuran, 98+%

CAS: 3208-16-0 | C6H8O | 96.13 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A15520.06
または、製品番号A15520-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3208-16-0
IUPAC Name2-ethylfuran
Molecular FormulaC6H8O
InChI KeyHLPIHRDZBHXTFJ-UHFFFAOYSA-N
SMILESCCC1=CC=CO1
さらに表示
Appearance (Color)Clear colorless to pale yellow
Transition Temperature1.4380-1.4420 @ 20?C
Assay (GC)>97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only