p-Toluoyl chloride, 99%
p-Toluoyl chloride, 99%
p-Toluoyl chloride, 99%
Thermo Scientific Chemicals

p-Toluoyl chloride, 99%

CAS: 874-60-2 | C8H7ClO | 154.593 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A15581.0E
または、製品番号A15581-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS874-60-2
IUPAC Name4-methylbenzoyl chloride
Molecular FormulaC8H7ClO
InChI KeyNQUVCRCCRXRJCK-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(Cl)=O
さらに表示
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5525-1.5555 @ 20?C
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

Solubility
Reacts with water alcohol.

Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
RUO – Research Use Only

General References:

  1. M H Karol; Y Alarie. Kaputsky. Antigens which detect IgE antibodies in workers sensitive to toluene diisocyanate. Clinical allergy. 1980,10 (1), 101-109.
  2. Shainaz M Landge; M Chidambaram; A.P Singh. Benzoylation of toluene with p-toluoyl chloride over triflic acid functionalized mesoporous Zr-TMS catalyst. Journal of Molecular Catalysis A: Chemical 2004, 213 (2), 257-266.