N-(3-Bromopropyl)phthalimide, 98%
N-(3-Bromopropyl)phthalimide, 98%
N-(3-Bromopropyl)phthalimide, 98%
N-(3-Bromopropyl)phthalimide, 98%
Thermo Scientific Chemicals

N-(3-Bromopropyl)phthalimide, 98%

CAS: 5460-29-7 | C11H10BrNO2 | 268.11 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15587.22
または、製品番号A15587-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5460-29-7
IUPAC Name2-(3-bromopropyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC11H10BrNO2
InChI KeyVKJCJJYNVIYVQR-UHFFFAOYSA-N
SMILESBrCCCN1C(=O)C2=CC=CC=C2C1=O
さらに表示
FormCrystalline powder or powder
Assay (GC)≥97.5%
CommentMay contain a small amount of dark particulates caused by trace bromine in the crystals
Identification (FTIR)Conforms
Melting Point (clear melt)70-76°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only