3-Bromoanisole, 98+%
3-Bromoanisole, 98+%
3-Bromoanisole, 98+%
Thermo Scientific Chemicals

3-Bromoanisole, 98+%

CAS: 2398-37-0 | C7H7BrO | 187.04 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15625.14
または、製品番号A15625-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2398-37-0
IUPAC Name1-bromo-3-methoxybenzene
Molecular FormulaC7H7BrO
InChI KeyPLDWAJLZAAHOGG-UHFFFAOYSA-N
SMILESCOC1=CC=CC(Br)=C1
さらに表示
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.5620-1.5660 @ 20?C
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. S. Ramalingam; S. Periandy. Spectroscopic investigation, computed IR intensity, Raman activity and vibrational frequency analysis on 3-bromoanisole using HF and DFT (LSDA/MPW1PW91) calculations. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2011, 78 (2), 835-843.
  2. Bobby Glover; Kimberly A. Harvey; Bing Liu; Matthew J. Sharp; and Maria F. Tymoschenko. Regioselective Palladium-Catalyzed Arylation of 3-Carboalkoxy Furan and Thiophene. Org. Lett. 2003, 5 (3), 301-304.