Thermo Scientific Chemicals

3-Methyl-2-cyclohexen-1-one, 98%, Thermo Scientific Chemicals

Catalog number: A15704.09
10 g, Each
Thermo Scientific Chemicals

3-Methyl-2-cyclohexen-1-one, 98%, Thermo Scientific Chemicals

Catalog number: A15704.09
10 g, Each
Quantity
Catalog number: A15704.09
also known as A15704-09
Price (USD)
Price: 75.50
Online price: 64.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1193-18-6
IUPAC Name
3-methylcyclohex-2-en-1-one
Molecular Formula
C7H10O
InChI Key
IITQJMYAYSNIMI-UHFFFAOYSA-N
SMILES
CC1=CC(=O)CCC1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4930-1.4970 @ 20?C

Description

3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text