Thermo Scientific Chemicals

3-Methylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals

Catalog number: A15772.14
25 g, Each
Thermo Scientific Chemicals

3-Methylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals

Catalog number: A15772.14
25 g, Each
Quantity
Catalog number: A15772.14
also known as A15772-14
Price (USD)
Price: 674.00
Online price: 572.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1455-18-1
IUPAC Name
3-methyl-1-benzothiophene
Molecular Formula
C9H8S
InChI Key
SEBRPHZZSLCDRQ-UHFFFAOYSA-N
SMILES
CC1=CSC2=CC=CC=C12
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥97.5%
Refractive Index
1.6225-1.6265 @ 20°C
Form
Liquid

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
participates in base-catalyzed Knoevenagel condensation reaction
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text