Thermo Scientific Chemicals

Brij|r L23, Thermo Scientific Chemicals

Catalog number: A15809.0I
5000 g, Each
Thermo Scientific Chemicals

Brij|r L23, Thermo Scientific Chemicals

Catalog number: A15809.0I
5000 g, Each
Quantity
Catalog number: A15809.0I
also known as A15809-0I
Price (USD)
Price: 270.00
Online price: 229.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9002-92-0
MDL Number
MFCD00043063
Acid Value
≤1.0 mg KOH/g
Colour (Hazen Units)
0-50
Appearance (Color)
White
Form
Waxy fused solid or pellets or flakes
Identification (FTIR)
Conforms

Description

Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (40 mg/ml) at 20°C methanol, chloroform and ethanol.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent. Used as a wetting agent, a detergent, or an emulsifier. Brij™L23 is a detergent used for measuring the concentration of carbon black in lung tissues.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text