2,6-Dibromo-4-isopropylaniline, 98%
2,6-Dibromo-4-isopropylaniline, 98%
2,6-Dibromo-4-isopropylaniline, 98%
Thermo Scientific Chemicals

2,6-Dibromo-4-isopropylaniline, 98%

CAS: 10546-65-3 | C9H11Br2N | 293.002 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A15932.22
または、製品番号A15932-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10546-65-3
IUPAC Name2,6-dibromo-4-(propan-2-yl)aniline
Molecular FormulaC9H11Br2N
InChI KeyCJEBZUFROMNDEK-UHFFFAOYSA-N
SMILESCC(C)C1=CC(Br)=C(N)C(Br)=C1
さらに表示
Appearance (Color)Cream or pale pink or pale green
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)43.5-49.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only