Thermo Scientific Chemicals

5-Methylbenzimidazole, 98%, Thermo Scientific Chemicals

Catalog number: A15957.22
100 g, Each
Thermo Scientific Chemicals

5-Methylbenzimidazole, 98%, Thermo Scientific Chemicals

Catalog number: A15957.22
100 g, Each
Quantity
Catalog number: A15957.22
also known as A15957-22
Price (USD)
Price: 592.00
Online price: 512.65
Your price:
Quantity
-

Chemical Identifiers

CAS
614-97-1
IUPAC Name
6-methyl-1H-1,3-benzodiazole
Molecular Formula
C8H8N2
InChI Key
RWXZXCZBMQPOBF-UHFFFAOYSA-N
SMILES
CC1=CC=C2N=CNC2=C1
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥97.5%
Form
Powder
Melting Point (clear melt)
110.0-116.0?C

Description

5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzimidazole is used as a camphor substitute. It is also used as fine chemical and medicine intermediate.

Solubility
Soluble in chloroform (25mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text