4-Fluorobenzeneboronic acid, 97%
4-Fluorobenzeneboronic acid, 97%
4-Fluorobenzeneboronic acid, 97%
Thermo Scientific Chemicals

4-Fluorobenzeneboronic acid, 97%

CAS: 1765-93-1 | C6H6BFO2 | 139.92 g/mol
製品番号(カタログ番号) A15991.03
または、製品番号A15991-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1765-93-1
IUPAC Name(4-fluorophenyl)boronic acid
Molecular FormulaC6H6BFO2
InChI KeyLBUNNMJLXWQQBY-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(F)C=C1
さらに表示
Appearance (Color)White to cream
FormPowder
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only