Thermo Scientific Chemicals

trans-3-Bromocinnamic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16035.22
100 g, Each
Thermo Scientific Chemicals

trans-3-Bromocinnamic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16035.22
100 g, Each
Quantity
Catalog number: A16035.22
also known as A16035-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14473-91-7
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%
Form
Powder
Melting Point (clear melt)
176.0-182.0?C

Description

trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Bromocinnamic acid is used as organic synthesis of raw materials, fine chemicals, pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text