Diethylene glycol mono-n-butyl ether, 99%
Diethylene glycol mono-n-butyl ether, 99%
Diethylene glycol mono-n-butyl ether, 99%
Thermo Scientific Chemicals

Diethylene glycol mono-n-butyl ether, 99%

CAS: 112-34-5 | C8H18O3 | 162.229 g/mol
数量:
1000 mL
5000 mL
10 L
製品番号(カタログ番号) A16068.AU
または、製品番号A16068-AU
価格(JPY)
-
数量:
1000 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS112-34-5
IUPAC Name2-(2-butoxyethoxy)ethan-1-ol
Molecular FormulaC8H18O3
InChI KeyOAYXUHPQHDHDDZ-UHFFFAOYSA-N
SMILESCCCCOCCOCCO
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Refractive Index1.4305-1.4335 @ 20?C
さらに表示
Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.

Solubility
Miscible with water, ethyl ether, ethanol acetone and benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents and light metals.
RUO – Research Use Only

General References:

  1. Wakoh, H.; Furuie, M.; Inaba, D.; Azuma, N.; Nakane, K.; Ogata, N.; Shimizu, T.; Ishimaru, O. Deep-colour vat dyeing of cotton knit fabric on a modified jet dyeing machine. Color. Technol. 2015, 131 (2), 136-141.
  2. Liu, L.; Kim, G. Y.; Hillier, A. C.; Chandra, A. Microstructural and electrochemical impedance study of nickel-Ce0.9Gd0.1O1.95 anodes for solid oxide fuel cells fabricated by ultrasonic spray pyrolysis. J. Power Sources 2011, 196 (6), 3026-3032.