Methyl 2-fluorobenzoate, 98%
Methyl 2-fluorobenzoate, 98%
Methyl 2-fluorobenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-fluorobenzoate, 98%

CAS: 394-35-4 | C8H7FO2 | 154.14 g/mol
製品番号(カタログ番号) A16133.14
または、製品番号A16133-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS394-35-4
IUPAC Namemethyl 2-fluorobenzoate
Molecular FormulaC8H7FO2
InChI KeyQAFJIJWLEBLXHH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1F
さらに表示
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.5000-1.5040 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only