2-Acetyl-5-methylfuran, 98+%
2-Acetyl-5-methylfuran, 98+%
2-Acetyl-5-methylfuran, 98+%
2-Acetyl-5-methylfuran, 98+%
Thermo Scientific Chemicals

2-Acetyl-5-methylfuran, 98+%

CAS: 1193-79-9 | C7H8O2 | 124.139 g/mol
数量:
10 g
25 g
50 g
製品番号(カタログ番号) A16172.14
または、製品番号A16172-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1193-79-9
IUPAC Name1-(5-methylfuran-2-yl)ethan-1-one
Molecular FormulaC7H8O2
InChI KeyKEFJLCGVTHRGAH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C)O1
さらに表示
Appearance (Color)Clear yellow
Identification (FTIR)Conforms
Refractive Index1.5115-1.5155 @ 20?C
FormLiquid
Assay (GC)≥98.0%

Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Keith Basey,; Jack G. Woolley. Alkaloids of Physalis alkekengi. Phytochemistry. 1973, 12 (10), 2557-2559.
  2. Liang Zhang,; Isao Kuniyoshi,; Mitsuyo Hirai,; Makoto Shoda. Oxidation of dimethyl sulfide byPseudomonas acidovorans DMR-11 isolated from peat biofilter. Biotechnology Letters. 1991, 13 (3), 223-228.