Formaldehyde sodium bisulfite, 95%
Formaldehyde sodium bisulfite, 95%
Formaldehyde sodium bisulfite, 95%
Thermo Scientific Chemicals

Formaldehyde sodium bisulfite, 95%

CAS: 870-72-4 | CH3NaO4S | 134.081 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A16194.0E
または、製品番号A16194-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS870-72-4
IUPAC Namesodium hydroxymethanesulfonate
Molecular FormulaCH3NaO4S
InChI KeyUOULCEYHQNCFFH-UHFFFAOYSA-M
SMILES[Na+].OCS([O-])(=O)=O
さらに表示
FormPowder
Assay (Titration)≥94.0 to ≤106.0%
Appearance (Color)White to pale cream
Formaldehyde sodium bisulfite, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It also acts as an pharmaceutical intermediate. It can effectively complex metal ion such as Cu, Zn, Pb, co-depositing with nickel ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formaldehyde sodium bisulfite, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It also acts as an pharmaceutical intermediate. It can effectively complex metal ion such as Cu, Zn, Pb, co-depositing with nickel ion.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic, Keep away from acids Strong oxidizing agents.
RUO – Research Use Only

General References:

  1. F Goldman.; H Yagoda. Collection and estimation of traces of formaldehyde in air. Ind. Eng. Chem. Anal. Ed.1943, 15, (6), 377-378.
  2. RR Miksch.; DW Anthon.; LZ Fanning. Modified pararosaniline method for the determination of formaldehyde in air. Anal. Chem.1981, 53, (13), 2118-2123.