Copper(II) acetate monohydrate, 98+%
Copper(II) acetate monohydrate, 98+%
Copper(II) acetate monohydrate, 98+%
Thermo Scientific Chemicals

Copper(II) acetate monohydrate, 98+%

CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol
数量:
250 g
1000 g
5000 g
製品番号(カタログ番号) A16203.0B
または、製品番号A16203-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6046-93-1
IUPAC Namecopper(2+) diacetate hydrate
Molecular FormulaC4H8CuO5
InChI KeyNWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILESO.[Cu++].CC([O-])=O.CC([O-])=O
さらに表示
Identification (FTIR)Conforms
Appearance (Color)Blue-green
FormCrystals or powder or crystalline powder
Assay (Iodometric Titration)≥98.0 to ≤102.0%
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nagababu, P.; Barui, A. K.; Thulasiram, B.; Devi, C. S.; Satyanarayana, S.; Patra, C. R.; Sreedhar, B. Antiangiogenic Activity of Mononuclear Copper (II) Polypyridyl Complexes for the Treatment of Cancers. J. Med. Chem. 2015, 58 (13), 5226-5241.
  2. Gogoi, K.; Deka, H.; Kumar, V.; Mondal, B. Oxo Transfer from Nitrogen Dioxide to Nitrito Group in a Copper(II) Complex. Inorg. Chem. 2015, 54 (10), 4799-4805.