Thermo Scientific Chemicals

Copper(II) acetate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A16203.30
250 g, Each
Thermo Scientific Chemicals

Copper(II) acetate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A16203.30
250 g, Each
Quantity
Catalog number: A16203.30
also known as A16203-30
Price (USD)
Price: 49.40
Online price: 41.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6046-93-1
IUPAC Name
copper(2+) diacetate hydrate
Molecular Formula
C4H8CuO5
InChI Key
NWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILES
O.[Cu++].CC([O-])=O.CC([O-])=O
Identification (FTIR)
Conforms
Appearance (Color)
Blue-green
Form
Crystals or powder or crystalline powder
Assay (Iodometric Titration)
≥98.0 to ≤102.0%

Description

Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text