D-Arginine monohydrochloride, 98%
D-Arginine monohydrochloride, 98%
D-Arginine monohydrochloride, 98%
Thermo Scientific Chemicals

D-Arginine monohydrochloride, 98%

CAS: 627-75-8 | C6H15ClN4O2 | 210.662 g/mol
製品番号(カタログ番号) A16222.14
または、製品番号A16222-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS627-75-8
IUPAC Namehydrogen (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Molecular FormulaC6H15ClN4O2
InChI KeyKWTQSFXGGICVPE-PGMHMLKASA-N
SMILES[H+].[Cl-].N[C@H](CCCN=C(N)N)C(O)=O
さらに表示
FormPowder
Optical Rotation-22? ? 1.5? (c=2 in 6M HCl)
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

Solubility
Soluble in water (50 mg/ml), and 3N HCl (5).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Sanford M. Birnbaum,; Milton Winitz,; Jesse P. Greenstein. A simplified preparation of d-arginine. Archives of Biochemistry and Biophysics. 1956 , 60 (2), 496-498.
  2. A. G. Eaton,; J. K. Doty. The Heat Production And Blood And Urine Constituents After Administration Of D-Arginine Monohydrochloride To The Dog. The Journal of Nutrition, 1939 .