Thermo Scientific Chemicals

Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: A16532.14
25 g, Each
Thermo Scientific Chemicals

Oxamic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: A16532.14
25 g, Each
Quantity
Catalog number: A16532.14
also known as A16532-14
Price (USD)
Price: 112.00
Online price: 95.65
Your price:
Quantity
-

Chemical Identifiers

CAS
565-73-1
IUPAC Name
sodium carbamoylformate
Molecular Formula
C2H2NNaO3
InChI Key
RQVZIJIQDCGIKI-UHFFFAOYSA-M
SMILES
[Na+].NC(=O)C([O-])=O
Appearance (Color)
White
Form
Powder
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.

Solubility
Soluble in water (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text