Thermo Scientific Chemicals

Lithium benzoate, 99+%, Thermo Scientific Chemicals

Catalog number: A16571.0B
1000 g, Each
Thermo Scientific Chemicals

Lithium benzoate, 99+%, Thermo Scientific Chemicals

Catalog number: A16571.0B
1000 g, Each
Quantity
Catalog number: A16571.0B
also known as A16571-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
553-54-8
IUPAC Name
lithium(1+) benzoate
Molecular Formula
C7H5LiO2
InChI Key
LDJNSLOKTFFLSL-UHFFFAOYSA-M
SMILES
[Li+].[O-]C(=O)C1=CC=CC=C1
Appearance (Color)
White to cream
Water Content (Karl Fischer Titration)
≤0.5%
Assay (Non-aqueous acid-base Titration)
≥99.0% to ≤101.0%
Form
Crystals or powder or crystalline powder

Description

In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

Solubility
Solubility in water is increased by sodium benzoate

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text