DL-Carnitine hydrochloride, 98+%
DL-Carnitine hydrochloride, 98+%
DL-Carnitine hydrochloride, 98+%
Thermo Scientific Chemicals

DL-Carnitine hydrochloride, 98+%

CAS: 461-05-2 | C7H17ClNO3+ | 198.667 g/mol
製品番号(カタログ番号) A16589.22
または、製品番号A16589-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS461-05-2
IUPAC Name(3-carboxy-2-hydroxypropyl)trimethylazanium chloride
Molecular FormulaC7H16ClNO3
InChI KeyJXXCENBLGFBQJM-UHFFFAOYNA-N
SMILES[Cl-].C[N+](C)(C)CC(O)CC(O)=O
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Melting point194°C to 200°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only