Thermo Scientific Chemicals

Methyl 2-amino-5-bromobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A16666.14
25 g, Each
Thermo Scientific Chemicals

Methyl 2-amino-5-bromobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A16666.14
25 g, Each
Quantity
Catalog number: A16666.14
also known as A16666-14
Price (USD)
Price: 126.00
Online price: 107.65
Your price:
Quantity
-

Chemical Identifiers

CAS
52727-57-8
Melting Point
68.0-74.0?C (clear to hazy)
Appearance (Color)
Pale cream to cream to pale gray or pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

Solubility
Soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents, strong reducing agents, acid halides, acid anhydrides, strong acids and strong bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text