2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
2,2':5',2″-Terthiophene, 99%
Thermo Scientific Chemicals

2,2':5',2″-Terthiophene, 99%

CAS: 1081-34-1 | C12H8S3 | 248.376 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) A16748.06
または、製品番号A16748-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1081-34-1
IUPAC Name5-(thiophen-2-yl)-2,2'-bithiophene
Molecular FormulaC12H8S3
InChI KeyKXSFECAJUBPPFE-UHFFFAOYSA-N
SMILESS1C=CC=C1C1=CC=C(S1)C1=CC=CS1
さらに表示
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)90.0-96.0?C
2,2?:5?,2??-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Murat Ates; Nuri Eren. Electrochemical copolymerization of carbazole and 2,2':5'-2″ terthiophene: characterization and micro-capacitor application. Iranian Polymer Journal. 2014, 23 581-589.
  2. Mohammad Shamsuddin Ahmed; Haesang Jeong; Jung-Min You; Seungwon Jeon. Synthesis and characterization of an electrochromic copolymer based on 2,2':5',2″-terthiophene and 3,4-ethylenedioxythiophene. Applied Nanoscience. 2012, 2 133-141.