L-(-)-Fucose, 98%
L-(-)-Fucose, 98%
L-(-)-Fucose, 98%
Thermo Scientific Chemicals

L-(-)-Fucose, 98%

CAS: 2438-80-4 | C6H12O5 | 164.16 g/mol
製品番号(カタログ番号) A16789.03
または、製品番号A16789-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2438-80-4
IUPAC Name(3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
Molecular FormulaC6H12O5
InChI KeySHZGCJCMOBCMKK-DHVFOXMCSA-N
SMILESC[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O
さらに表示
Water Content (Karl Fischer Titration)≤1.0% (non-U.S. specification)
Appearance (Color)White to pale cream
FormPowder or crystalline powder or lumps
Assay (HPLC)≥97.5%
Assay from Suppliers CofA≥97.5% (HPLC, U.S. specification)
さらに表示
L-(-)-Fucose finds application in cosmetics, pharmaceuticals and dietary supplements. It is used in the determination of antigen in A and B blood group. It is also used in the selection-mediated leukocyte-endothelial adhesion and host-microbe interactions. It is also used in anti aging creams as well as to promote the accelerated healing of wounds and to reduce allergy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Fucose finds application in cosmetics, pharmaceuticals and dietary supplements. It is used in the determination of antigen in A and B blood group. It is also used in the selection-mediated leukocyte-endothelial adhesion and host-microbe interactions. It is also used in anti aging creams as well as to promote the accelerated healing of wounds and to reduce allergy.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

General References:

  1. Becerra, J. E.; Yebra, M. J.; Monedero, V. An L-Fucose Operon in the Probiotic Lactobacillus rhamnosus GG Is Involved in Adaptation to Gastrointestinal Conditions. Appl. Environ. Microbiol. 2015, 81 (11), 3880-3888.
  2. Liu, Z.; Yoshihara, A.; Wormald, M. R.; Jenkinson, S. F.; Gibson, V.; Izumori, K.; Fleet, G. W. J. L-Fucose from Vitamin C with Only Acetonide Protection. Org. Lett. 2014, 16 (21), 5663-5665.