Thermo Scientific Chemicals

D-Gluconic acid, 50% aq. soln., Thermo Scientific Chemicals

Catalog number: A16806.0F
2500 mL, Each
Thermo Scientific Chemicals

D-Gluconic acid, 50% aq. soln., Thermo Scientific Chemicals

Catalog number: A16806.0F
2500 mL, Each
Quantity
Catalog number: A16806.0F
also known as A16806-0F
Price (USD)
Price: 80.70
Online price: 68.65
Your price:
Quantity
-

Chemical Identifiers

CAS
526-95-4
IUPAC Name
potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular Formula
C6H11KO7
InChI Key
HLCFGWHYROZGBI-JJKGCWMISA-M
SMILES
[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Appearance (Color)
Clear yellow
Form
Liquid
Acid-base back titration
≥48 to ≤54% w/w
Identification (FTIR)
Conforms
Optical Rotation
+10 ? 2? (neat)

Description

Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text