3-Chloro-1-propanol, 98%, stab.
3-Chloro-1-propanol, 98%, stab.
3-Chloro-1-propanol, 98%, stab.
Thermo Scientific Chemicals

3-Chloro-1-propanol, 98%, stab.

CAS: 627-30-5 | C3H7ClO | 94.538 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A16871.14
または、製品番号A16871-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS627-30-5
IUPAC Name3-chloropropan-1-ol
Molecular FormulaC3H7ClO
InChI KeyLAMUXTNQCICZQX-UHFFFAOYSA-N
SMILESOCCCCl
さらに表示
Appearance (Color)Clear colorless to yellow
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Stabilizer0.1% Na2CO3
さらに表示
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 20°C 300g/L.

Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Michael J. Fuller.; E.Bright Wilson.; Walther Caminatia. Rotational isomerism in 3-chloro-1-propanol from the microwave spectrum.J. Mol. Spectrosc.1982, 96(1), 131-145.
  2. N. Baggett.' Md. Mosihuzzaman.; J.M. Webber. Benzylidenation in basic medium: the reation of methyl 2,3-di-O-methyl-6-O-toluene-p-sulphonyl-α-d-glucopyranoside and 3-chloro-1-propanol with benzaldehyde.Carbohydr. Res.1969, 11(2), 263-266.