4-(Trifluoromethyl)benzyl bromide, 98%
4-(Trifluoromethyl)benzyl bromide, 98%
4-(Trifluoromethyl)benzyl bromide, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl bromide, 98%

CAS: 402-49-3 | C8H6BrF3 | 239.035 g/mol
製品番号(カタログ番号) A16895.06
または、製品番号A16895-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS402-49-3
IUPAC Name1-(bromomethyl)-4-(trifluoromethyl)benzene
Molecular FormulaC8H6BrF3
InChI KeyIKSNDOVDVVPSMA-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(CBr)C=C1
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Refractive Index1.4895-1.4935 @ 20?C
Appearance (Color)White to cream
Assay (GC)≥97.5%
さらに表示
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
M. Karnan; V. Balachandran; M. Murugan; M.K. Murali; A. Nataraj. Vibrational (FT-IR and FT-Raman) spectra, NBO, HOMO-LUMO, Molecular electrostatic potential surface and computational analysis of 4-(trifluoromethyl)benzylbromide. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy.2013, 116 84-95.