4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%
Thermo Scientific Chemicals

4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%

CAS: 315-30-0 | C5H4N4O | 136.11 g/mol
製品番号(カタログ番号) A16974.14
または、製品番号A16974-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS315-30-0
IUPAC Name1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular FormulaC5H4N4O
InChI KeyOFCNXPDARWKPPY-UHFFFAOYSA-N
SMILESO=C1N=CN=C2NNC=C12
さらに表示
Assay (HPLC)≥97.5%
Appearance (Color)White
FormPowder
Xanthine oxidase inhibitor; decreases uric acid production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only