Dibromoacetonitrile, 94%
Dibromoacetonitrile, 94%
Dibromoacetonitrile, 94%
Thermo Scientific Chemicals

Dibromoacetonitrile, 94%

CAS: 3252-43-5 | C2HBr2N | 198.845 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A16994.06
または、製品番号A16994-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3252-43-5
IUPAC Name2,2-dibromoacetonitrile
Molecular FormulaC2HBr2N
InChI KeyNDSBDLSWTGLNQA-UHFFFAOYSA-N
SMILESBrC(Br)C#N
さらに表示
Assay (GC)≥92.0%
FormPowder or clear liquid as melt
Appearance (Color)Colorless to yellow to orange
Refractive Index1.5385-1.5455 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only