1-Phenylpiperazine hydrochloride, 99%
1-Phenylpiperazine hydrochloride, 99%
1-Phenylpiperazine hydrochloride, 99%
Thermo Scientific Chemicals

1-Phenylpiperazine hydrochloride, 99%

CAS: 2210-93-7 | C10H15ClN2 | 198.694 g/mol
製品番号(カタログ番号) A16995.22
または、製品番号A16995-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2210-93-7
IUPAC Namehydrogen 1-phenylpiperazine chloride
Molecular FormulaC10H15ClN2
InChI KeyNQNZNONJZASOKL-UHFFFAOYSA-N
SMILES[H+].[Cl-].C1CN(CCN1)C1=CC=CC=C1
さらに表示
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or pink or grey
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Assay (Freebase GC)≥98.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only