Ethyl 2-thiouracil-5-carboxylate, 99%
Ethyl 2-thiouracil-5-carboxylate, 99%
Ethyl 2-thiouracil-5-carboxylate, 99%
Ethyl 2-thiouracil-5-carboxylate, 99%
Thermo Scientific Chemicals

Ethyl 2-thiouracil-5-carboxylate, 99%

CAS: 38026-46-9 | C7H8N2O3S | 200.21 g/mol
製品番号(カタログ番号) A17046.14
または、製品番号A17046-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS38026-46-9
IUPAC Nameethyl 4-oxo-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Molecular FormulaC7H8N2O3S
InChI KeyFQFSHLBWRUOCPX-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CNC(=S)NC1=O
さらに表示
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only