2,2-Dimethylbutyric acid, 97%
2,2-Dimethylbutyric acid, 97%
2,2-Dimethylbutyric acid, 97%
Thermo Scientific Chemicals

2,2-Dimethylbutyric acid, 97%

CAS: 595-37-9 | C6H12O2 | 116.16 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A17072.18
または、製品番号A17072-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS595-37-9
IUPAC Name2,2-dimethylbutanoic acid
Molecular FormulaC6H12O2
InChI KeyVUAXHMVRKOTJKP-UHFFFAOYSA-N
SMILESCCC(C)(C)C(O)=O
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Refractive Index1.4125-1.4175 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only