1,4-Di-tert-butylbenzene, 98%
1,4-Di-tert-butylbenzene, 98%
1,4-Di-tert-butylbenzene, 98%
Thermo Scientific Chemicals

1,4-Di-tert-butylbenzene, 98%

CAS: 1012-72-2 | C14H22 | 190.33 g/mol
製品番号(カタログ番号) A17146.14
または、製品番号A17146-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1012-72-2
IUPAC Name1,4-di-tert-butylbenzene
Molecular FormulaC14H22
InChI KeyOOWNNCMFKFBNOF-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)C(C)(C)C
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)74.0-80.0?C
Appearance (Color)White
1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.

Solubility
Insoluble in water. Soluble in ethanol, benzene, carbon tetrachloride.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. GH Penner.; JM Polson.; C Stuart. Deuterium NMR and x-ray crystallographic studies of guest and host motions in the thiourea/1,4-di-tert-butylbenzene inclusion compound. J. Phys. Chem. 199296 (12), 5121-5129.
  2. N Gart.; CL Leci.; S Sarig. The effect of solvents on crystal habit of 1, 4-di-tert-butylbenzene (DTBB). Journal of Crystal Growth. 198154 (2), 227-231.