MOPS sodium salt, 97%
MOPS sodium salt, 97%
MOPS sodium salt, 97%
Thermo Scientific Chemicals

MOPS sodium salt, 97%

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol
製品番号(カタログ番号) A17214.22
または、製品番号A17214-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS71119-22-7
IUPAC Namesodium 3-(morpholin-4-yl)propane-1-sulfonate
Molecular FormulaC7H14NNaO4S
InChI KeyMWEMXEWFLIDTSJ-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCCN1CCOCC1
さらに表示
Identification (FTIR)Conforms (non-U.S. specification)
Acid-base back titration≥96.0 to ≤104.0% (non-U.S. specification)
Assay from Suppliers CofA≥96.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Appearance (Color)White to pale cream
さらに表示
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

Solubility
Soluble in water (523 g/L at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Munessar Sankar.; Roger G. Bates. Buffers for the physiological pH range: thermodynamic constants of 3-(N-morpholino)propanesulfonic acid from 5 to 50.degree.C.Anal. Chem.,1978,50(13), 1922-1924.
  2. K M Pan.; M Baldwin.; J Nguyen.; M Gasset.; A Serban.; D Groth.; I Mehlhorn.; Z Huang.; R J Fletterick.; and F E Cohen. Conversion of alpha-helices into beta-sheets features in the formation of the scrapie prion proteins.Proc. Natl. Acad. Sci.1993,90,(23), 10962-10966.